Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RanGAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273649.1 | NP_001260578.1 | 2 - 3 | 925 | 60 | |
NM_001273650.1 | NP_001260579.1 | 3 - 4 | 1061 | 60 | ||
NM_057364.4 | NP_476712.1 | 3 - 4 | 1318 | 60 | ||
GenBank mRNA | AY061219.1 | - | 2 - 3 | 910 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |