Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sli | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103863.3 | NP_001097333.1 | 2 - 3 | 1468 | 79 | |
NM_001103864.3 | NP_001097334.1 | 1 - 2 | 625 | 79 | ||
NM_001274088.1 | NP_001261017.1 | 2 - 3 | 1468 | 79 | ||
NM_057379.4 | NP_476727.1 | 2 - 3 | 1468 | 79 | ||
NM_057380.4 | NP_476728.1 | 2 - 3 | 1468 | 79 | ||
NM_057381.4 | NP_476729.1 | 3 - 4 | 1530 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |