Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057382.3 | NP_476730.1 | 1 | 1364 | 84 | |
GenBank mRNA | AY047536.1 | - | 1 | 1347 | 84 | |
X66096.1 | - | 1377 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |