Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sog | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272647.2 | NP_001259576.1 | 6 - 7 | 4051 | 56 | |
NM_001272649.2 | NP_001259578.1 | 5 - 6 | 3733 | 56 | ||
NM_057388.4 | NP_476736.1 | 5 - 6 | 3733 | 56 | ||
GenBank mRNA | BT053679.1 | - | 6 - 7 | 4051 | 56 | |
U18774.1 | - | 5 - 6 | 4078 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |