Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tud | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299736.1 | NP_001286665.1 | 3 - 4 | 1367 | 68 | |
NM_057425.3 | NP_476773.1 | 3 - 4 | 1367 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |