Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tup | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299139.1 | NP_001286068.1 | 6 - 7 | 1403 | 138 | |
NM_057426.4 | NP_476774.1 | 6 - 7 | 1403 | 138 | ||
NM_057427.3 | NP_476775.1 | 6 - 7 | 1403 | 138 | ||
GenBank mRNA | AF145674.1 | - | 6 - 7 | 1388 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |