Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Axs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057429.3 | NP_476777.1 | 3 - 4 | 734 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |