Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Axs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057429.3 | NP_476777.1 | 7 - 8 | 2364 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster Inventoried |