Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ush | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272899.1 | NP_001259828.1 | 3 - 4 | 1506 | 70 | |
NM_001272900.1 | NP_001259829.1 | 3 - 4 | 1306 | 70 | ||
NM_001272901.1 | NP_001259830.1 | 3 - 4 | 1094 | 70 | ||
NM_001272902.1 | NP_001259831.1 | 4 - 5 | 2936 | 70 | ||
NM_057432.3 | NP_476780.1 | 3 - 4 | 1576 | 70 | ||
GenBank mRNA | BT015278.1 | - | 3 - 4 | 1289 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |