Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vinc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297884.1 | NP_001284813.1 | 4 - 5 | 2563 | 99 | |
NM_001297885.1 | NP_001284814.1 | 4 - 5 | 2657 | 99 | ||
NM_057472.3 | NP_476820.1 | 4 - 5 | 2585 | 99 | ||
GenBank mRNA | AY128501.1 | - | 4 - 5 | 2515 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |