Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273384.2 | NP_001260313.1 | 3 - 4 | 793 | 74 | |
NM_057489.5 | NP_476837.1 | 3 - 4 | 793 | 74 | ||
GenBank mRNA | AY051981.1 | - | 3 - 4 | 773 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |