Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: U2af50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258779.1 | NP_001245708.1 | 2 - 3 | 608 | 79 | |
NM_001258780.1 | NP_001245709.1 | 2 - 3 | 635 | 79 | ||
NM_001258781.2 | NP_001245710.1 | 2 - 3 | 737 | 79 | ||
NM_057543.4 | NP_476891.1 | 2 - 3 | 791 | 79 | ||
GenBank mRNA | AY069320.1 | - | 2 - 3 | 701 | 79 | |
L23404.1 | - | 2 - 3 | 702 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |