Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dbp45A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057579.5 | NP_476927.1 | 1 - 2 | 155 | 69 | |
GenBank mRNA | AY058728.1 | - | 1 - 2 | 128 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |