Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dbp45A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057579.5 | NP_476927.1 | 4 - 5 | 1225 | 85 | |
GenBank mRNA | AY058728.1 | - | 4 - 5 | 1198 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |