Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014462.3 | NP_001014462.1 | 6 - 7 | 1582 | 59 | |
NM_001273045.1 | NP_001259974.1 | 6 - 7 | 1579 | 59 | ||
NM_001298640.1 | NP_001285569.1 | 5 - 6 | 1376 | 59 | ||
NM_057588.3 | NP_476936.3 | 5 - 6 | 1667 | 59 | ||
NM_057589.3 | NP_476937.3 | 6 - 7 | 1682 | 59 | ||
NM_134297.5 | NP_599124.1 | 7 - 8 | 1598 | 59 | ||
NM_134298.5 | NP_599125.1 | 6 - 7 | 1584 | 59 | ||
NM_134299.5 | NP_599126.1 | 6 - 7 | 1596 | 59 | ||
NM_134300.4 | NP_599127.1 | 7 - 8 | 1614 | 59 | ||
GenBank mRNA | BT011197.1 | - | 6 - 7 | 1557 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |