Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TfIIS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273528.1 | NP_001260457.1 | 1 - 2 | 319 | 97 | |
NM_057619.4 | NP_476967.1 | 1 - 2 | 319 | 97 | ||
GenBank mRNA | AY051843.1 | - | 1 - 2 | 233 | 97 | |
X53670.1 | - | 296 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |