Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wnt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273258.2 | NP_001260187.1 | 1 - 2 | 1810 | 80 | |
NM_057624.4 | NP_476972.2 | 1 - 2 | 1810 | 80 | ||
GenBank mRNA | BT120121.1 | - | 1 - 2 | 1806 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |