Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: poe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273293.1 | NP_001260222.1 | 2 - 3 | 275 | 65 | |
NM_057638.4 | NP_476986.1 | 2 - 3 | 275 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |