Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299624.1 | NP_001286553.1 | 11 - 12 | 1683 | 76 | |
NM_057661.3 | NP_477009.1 | 11 - 12 | 1686 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |