Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tig | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298741.1 | NP_001285670.1 | 5 - 6 | 6705 | 59 | |
NM_057685.4 | NP_477033.1 | 5 - 6 | 6705 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |