Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wee1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273238.1 | NP_001260167.1 | 2 - 3 | 1053 | 51 | |
NM_057687.4 | NP_477035.1 | 2 - 3 | 1053 | 51 | ||
GenBank mRNA | AY118942.1 | - | 2 - 3 | 999 | 51 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |