Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Idgf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299570.1 | NP_001286499.1 | 2 - 3 | 470 | 65 | |
NM_057733.4 | NP_477081.1 | 1 - 2 | 200 | 65 | ||
GenBank mRNA | AY058510.1 | - | 1 - 2 | 170 | 65 | |
BT029919.1 | - | 1148 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |