Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273803.1 | NP_001260732.1 | 3 - 4 | 687 | 70 | |
NM_057742.4 | NP_477090.1 | 3 - 4 | 687 | 70 | ||
GenBank mRNA | BT003315.1 | - | 3 - 4 | 641 | 70 | |
D84313.1 | - | 3 - 4 | 639 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |