Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Diap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057779.5 | NP_477127.1 | 2 - 3 | 1559 | 82 | |
NM_176182.2 | NP_788362.1 | 3 - 4 | 1450 | 82 | ||
GenBank mRNA | AY051844.1 | - | 2 - 3 | 1528 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |