Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pi3K59F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057785.5 | NP_477133.1 | 4 - 5 | 1847 | 67 | |
GenBank mRNA | AY058362.1 | - | 4 - 5 | 1824 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |