Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srp19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057787.5 | NP_477135.2 | 1 - 2 | 178 | 83 | |
GenBank mRNA | AY089689.1 | - | 1 - 2 | 159 | 83 | |
KX531857.1 | - | 1 - 2 | 65 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |