Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unpg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057798.3 | NP_477146.1 | 2 - 3 | 1209 | 69 | |
GenBank mRNA | BT022753.1 | - | 2 - 3 | 1415 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |