Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lic | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057814.4 | NP_477162.1 | 3 - 4 | 921 | 71 | |
GenBank mRNA | AF035550.1 | - | 3 - 4 | 843 | 71 | |
AF041136.1 | - | 3 - 4 | 908 | 71 | ||
AY052103.1 | - | 3 - 4 | 903 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |