Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057884.5 | NP_477232.1 | 3 - 4 | 1080 | 89 | |
GenBank mRNA | AF043263.1 | - | 3 - 4 | 1042 | 89 | |
BT044099.1 | - | 3 - 4 | 1079 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |