Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dve | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057893.5 | NP_477241.2 | 2 - 3 | 243 | 61 | |
NM_057894.4 | NP_477242.1 | 3 - 4 | 1677 | 61 | ||
GenBank mRNA | BT025086.1 | - | 3 - 4 | 1662 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |