Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM4SF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038879.2 | NP_001033968.1 | 3 - 4 | 1691 | 57 | |
NM_001274230.1 | NP_001261159.1 | 4 - 5 | 1336 | 57 | ||
NM_057900.4 | NP_477248.1 | 4 - 5 | 1026 | 57 | ||
GenBank mRNA | BT072819.1 | - | 4 - 5 | 800 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |