Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sdk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169159.2 | NP_001162630.1 | 7 - 8 | 2497 | 56 | |
NM_001169160.2 | NP_001162631.1 | 8 - 9 | 1865 | 56 | ||
NM_001297827.1 | NP_001284756.1 | 9 - 10 | 2134 | 56 | ||
NM_001297828.1 | NP_001284757.1 | 8 - 9 | 1999 | 56 | ||
NM_057941.5 | NP_477289.5 | 8 - 9 | 1934 | 56 | ||
NM_057942.5 | NP_477290.6 | 8 - 9 | 2051 | 56 | ||
NM_134314.4 | NP_599141.5 | 8 - 9 | 1999 | 56 | ||
NM_134315.4 | NP_599142.5 | 8 - 9 | 2051 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |