Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nle | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057946.4 | NP_477294.2 | 1 - 2 | 1032 | 70 | |
GenBank mRNA | AY089286.1 | - | 1 - 2 | 1074 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |