Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rnh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057967.5 | NP_477315.1 | 2 - 3 | 427 | 71 | |
GenBank mRNA | AY089374.1 | - | 2 - 3 | 433 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |