Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mct1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297881.1 | NP_001284810.1 | 6 - 7 | 1601 | 78 | |
NM_057986.4 | NP_477334.2 | 6 - 7 | 1818 | 78 | ||
GenBank mRNA | BT010002.1 | - | 6 - 7 | 1816 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |