Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hs2st | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057991.5 | NP_477339.1 | 4 - 5 | 935 | 89 | |
GenBank mRNA | AY058422.1 | - | 4 - 5 | 946 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |