Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103847.2 | NP_001097317.1 | 4 - 5 | 858 | 65 | |
NM_001274051.1 | NP_001260980.1 | 3 - 4 | 527 | 65 | ||
NM_080074.4 | NP_524813.2 | 3 - 4 | 979 | 65 | ||
NM_137146.3 | NP_610990.2 | 3 - 4 | 979 | 65 | ||
NM_166070.3 | NP_725419.2 | 3 - 4 | 979 | 65 | ||
GenBank mRNA | BT150395.1 | - | 3 - 4 | 977 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |