Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080131.5 | NP_524870.1 | 1 - 2 | 446 | 57 | |
GenBank mRNA | AF221546.2 | - | 1 - 2 | 560 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |