Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080136.3 | NP_524875.1 | 3 - 4 | 673 | 105 | |
GenBank mRNA | AF239214.1 | - | 3 - 4 | 690 | 105 | |
BT004504.1 | - | 3 - 4 | 690 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |