Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klp54D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259469.2 | NP_001246398.1 | 1 - 2 | 503 | 90 | |
NM_001299610.1 | NP_001286539.1 | 1 - 2 | 503 | 90 | ||
NM_001316425.1 | NP_001303354.1 | 1 - 2 | 503 | 90 | ||
NM_001316426.1 | NP_001303355.1 | 1 - 2 | 503 | 90 | ||
NM_080144.3 | NP_524883.2 | 1 - 2 | 503 | 90 | ||
GenBank mRNA | BT120104.1 | - | 1 - 2 | 479 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Drosophila melanogaster |