Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klp54D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259469.2 | NP_001246398.1 | 2 - 3 | 676 | 68 | |
NM_001299610.1 | NP_001286539.1 | 2 - 3 | 676 | 68 | ||
NM_001316425.1 | NP_001303354.1 | 2 - 3 | 676 | 68 | ||
NM_001316426.1 | NP_001303355.1 | 2 - 3 | 676 | 68 | ||
NM_080144.3 | NP_524883.2 | 2 - 3 | 676 | 68 | ||
GenBank mRNA | BT120104.1 | - | 2 - 3 | 652 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |