Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eip63F-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202116.1 | NP_001189045.1 | 3 - 4 | 461 | 72 | |
NM_001202117.2 | NP_001189046.1 | 5 - 6 | 506 | 72 | ||
NM_001202118.2 | NP_001189047.1 | 4 - 5 | 686 | 72 | ||
NM_080163.4 | NP_524902.2 | 5 - 6 | 765 | 72 | ||
GenBank mRNA | BT088940.1 | - | 5 - 6 | 744 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |