Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsp60B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080186.4 | NP_524925.1 | 1 | 2099 | 61 | |
GenBank mRNA | BT001426.1 | - | 2027 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |