Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wek | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273543.1 | NP_001260472.1 | 1 - 2 | 248 | 83 | |
NM_080191.3 | NP_524930.1 | 1 - 2 | 248 | 83 | ||
GenBank mRNA | AY051673.1 | - | 1 - 2 | 216 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |