Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cmet | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273451.1 | NP_001260380.1 | 7 - 8 | 5611 | 69 | |
NM_001273453.1 | NP_001260382.1 | 7 - 8 | 5611 | 69 | ||
NM_080254.3 | NP_524993.3 | 8 - 9 | 5776 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |