Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080264.4 | NP_525003.2 | 1 - 2 | 274 | 81 | |
GenBank mRNA | AY071378.1 | - | 1 - 2 | 236 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |