Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgcl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080291.4 | NP_525030.1 | 1 | 1206 | 79 | |
GenBank mRNA | BT030803.1 | - | 1 | 1200 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |