Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scgdelta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169168.2 | NP_001162639.2 | 4 - 5 | 1320 | 81 | |
NM_001169169.2 | NP_001162640.1 | 4 - 5 | 1314 | 81 | ||
NM_001169170.2 | NP_001162641.1 | 3 - 4 | 1507 | 81 | ||
NM_080300.3 | NP_525039.1 | 4 - 5 | 1320 | 81 | ||
GenBank mRNA | AF277393.1 | - | 3 - 4 | 1065 | 81 | |
BT064640.1 | - | 4 - 5 | 1312 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |