Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4ae1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080305.4 | NP_525044.1 | 2 - 3 | 705 | 77 | |
GenBank mRNA | AY058450.1 | - | 2 - 3 | 705 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |