Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: per | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272265.1 | NP_001259194.1 | 3 - 4 | 1505 | 76 | |
NM_080317.2 | NP_525056.2 | 3 - 4 | 1505 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |